2-(Diallylamino)ethylamine ≥97%

Supplier: Matrix Scientific

007545-5G 007545-25G
101779-600EA 358.46 USD
101779-600 101779-598
2-(Diallylamino)ethylamine ≥97%
2-(Diallylamino)ethylamine

Hazard: Corrosive/Irritant

Formula: C₈H₁₆N₂
Boiling Pt: 79 °C (20 Torr)
Density: 0.8523
MDL Number: MFCD03452794
CAS Number: 25077-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR