N-Butyl-N-propylamine ≥97%

Supplier: Matrix Scientific

007527-5G 007527-25G
101779-584EA 209.23 USD
101779-584 101779-582
N-Butyl-N-propylamine ≥97%
N-Butyl-N-propylamine

Hazard: Corrosive/Irritant

Formula: C₇H₁₇N
Boiling Pt: 134 °C
Density: 0.7497
MDL Number: MFCD00015232
CAS Number: 20193-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR