N-Ethyl-N-butylethylenediamine

Supplier: Matrix Scientific

007231-5G
101779-526EA 92.31 USD
101779-526
N-Ethyl-N-butylethylenediamine
N-Ethyl-N-butylethylenediamine

Hazard: Irritant-Harmful

Formula: C₈H₂₀N₂
Boiling Pt: 165…167 °C
Density: 0.8197
MDL Number: MFCD00047953
CAS Number: 19435-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR