2-(N-Methyl-N-butylamino)ethylamine
Supplier: Matrix Scientific
007227-10G
101779-516EA
138.46
USD
101779-516
2-(N-Methyl-N-butylamino)ethylamine
2-(N-Methyl-N-butylamino)ethylamine
Hazard: Corrosive
Formula:
C₇H₁₈N₂ Boiling Pt: 150 °C |
MDL Number:
MFCD00047952 CAS Number: 15404-06-5 |
Specification Test Results
Boiling point | 150° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...