5-(Diisobutylamino)amylamine

Supplier: Matrix Scientific

007225-5G 007225-25G
101779-514EA 412.31 USD
101779-514 101779-512
5-(Diisobutylamino)amylamine
5-(Diisobutylamino)amylamine

Hazard: Corrosive

Formula: C₁₃H₃₀N₂
Boiling Pt: 95 °C ( 4 mmHg)
MDL Number: MFCD00048343
CAS Number: 209803-39-4

Order Now

Specification Test Results

Boiling point 95°/ 4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR