5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride ≥97%

Supplier: Matrix Scientific

007151-1G 007151-250MG
101779-418EA 92.31 USD
101779-418 101779-420
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride ≥97%
5-[1-Methyl-3-(trifluoromethyl)pyrazole-5-yl]thiophene-2-sulfonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₉H₆ClF₃N₂O₂S₂
MW: 330.74 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD00830811
CAS Number: 230295-11-1

Order Now

Specification Test Results

Melting point 65-67°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR