Fluoren-9-one-2,7-disulfonyl chloride ≥97%

Supplier: Matrix Scientific

007146-1G
101779-410EA 526.15 USD
101779-410
Fluoren-9-one-2,7-disulfonyl chloride ≥97%
Fluoren-9-one-2,7-disulfonyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₁₃H₆Cl₂O₅S₂
Melting Pt: 209…212 °C
MDL Number: MFCD00218830
CAS Number: 13354-21-7

Order Now

Specification Test Results

Melting point 209-212°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR