4-Ethylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Ethylbenzene-1-sulfonyl chloride4-Ethylbenzene-1-sulphonyl chloride

007143-5G
101779-408EA 55.38 USD
101779-408
4-Ethylbenzenesulfonyl chloride ≥97%
4-Ethylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₈H₉ClO₂S
MW: 204.68 g/mol
Melting Pt: 12 °C
Density: 1.268
MDL Number: MFCD00041480
CAS Number: 16712-69-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Melting point 12°
Density 1.268
ND20 1.544

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR