2-Chlorothiophene-3,5-bis(sulfonyl chloride) ≥97%
Supplier: Matrix Scientific
007137-5G
007137-1G
101779-382EA
213.85
USD
101779-382
101779-384
2-Chlorothiophene-3,5-bis(sulfonyl chloride) ≥97%
2-Chlorothiophene-3,5-bis(sulfonyl chloride)
Hazard: Corrosive/Moisture Sensitive
Formula:
C₄HCl₃O₄S₃ Melting Pt: 79…84 °C |
MDL Number:
MFCD00052154 CAS Number: 86806-70-4 |
Specification Test Results
Melting point | 79-84° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...