3-Chloro-2-methylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-methylbenzene-1-sulfonyl chloride3-Chloro-2-methylbenzene-1-sulphonyl chloride

007134-5G 007134-25G
101779-376EA 90.77 USD
101779-376 101779-374
3-Chloro-2-methylbenzenesulfonyl chloride ≥97%
3-Chloro-2-methylbenzenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₇H₆Cl₂O₂S
MW: 225.09 g/mol
Melting Pt: 70…72 °C
MDL Number: MFCD00052198
CAS Number: 80563-86-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 70-72°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR