3-Bromo-2-chloro-5-thiophenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-5-chlorothiophene-2-sulphonyl chloride4-Bromo-5-chlorothiophene-2-sulfonyl chloride

007127-1G
101779-352EA 186.15 USD
101779-352
3-Bromo-2-chloro-5-thiophenesulfonyl chloride ≥97%
3-Bromo-2-chloro-5-thiophenesulphonyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₄HBrCl₂O₂S₂
MW: 295.9895 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD00052094
CAS Number: 166964-35-8

Order Now

Specification Test Results

Melting point 63-65°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR