3-Fluoropyridine ≥99%

Supplier: Matrix Scientific

006670-100G
101779-212EA 63.08 USD
101779-212
3-Fluoropyridine ≥99%
3-Fluoropyridine

Hazard: Flammable/Irritant

Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 107…108 °C
Density: 1.13
MDL Number: MFCD00006374
CAS Number: 372-47-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 107-108°
Density 1.13
ND20 1.466

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR