Cyclopropylacetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Cyclopropylacetonitrile
006619-25G
006619-5G
101779-182EA
204.62
USD
101779-182
101779-180
Cyclopropylacetonitrile ≥95%
Cyclopropylacetonitrile
Hazard: Flammable/Toxic/Lachrymator
Formula:
C₅H₇N MW: 81.12 g/mol Boiling Pt: 142…144 °C Density: 0.89 Flash Pt: 43 °C |
MDL Number:
MFCD00041523 CAS Number: 6542-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...