Bromocyclopropane ≥99%
Supplier: Matrix Scientific
Synonyms:
Cyclopropyl bromide
006592-500G
006592-100G
102550-874EA
469.23
USD
102550-874
101779-164
Bromocyclopropane ≥99%
Bromocyclopropane
TCA listed: T
Hazard: Flammable/Refrigerate/Lachrymator
Formula:
C₃H₅Br MW: 120.98 g/mol Boiling Pt: 69 °C Density: 1.51 |
MDL Number:
MFCD00001271 CAS Number: 4333-56-6 UN: 1993 ADR: 3,II |
Specification Test Results
Boiling point | 69° |
Density | 1.51 |
ND20 | 1.46 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...