Ethyl-4,4,4-trifluoro-3-hydroxybutyrate ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4,4-Trifluoro-3-hydroxybutyric Acid Ethyl EsterEthyl 4,4,4-trifluoro-3-hydroxybutyrate

006587-25G
101779-160EA 36.92 USD
101779-160
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate ≥97%
Ethyl-4,4,4-trifluoro-3-hydroxybutyrate

Purity: 97%
CAS: 372-30-5
MDL Number: MFCD00040842
Molecular Formula: C6H9F3O3
Molecular Weight: 186,13
Boiling Point: 90-91°C/50mm
Hazard: Irritant

Formula: C₆H₉F₃O₃
MW: 186.13 g/mol
Boiling Pt: 90…91 °C (50 mmHg)
Melting Pt: 23 °C
MDL Number: MFCD00040842
CAS Number: 372-30-5

Order Now

Specification Test Results

Boiling point 90-91°/50m
ND20 1.374

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR