1-Fluorobutane ≥97%

Supplier: Matrix Scientific

006511-10G
101779-140EA 444.62 USD
101779-140
1-Fluorobutane ≥97%
1-Fluorobutane

TCA listed: T
Hazard: Flammable

Formula: C₄H₉F
MW: 76.11 g/mol
Boiling Pt: 31.9 °C (745 mmHg)
Density: 0.7761
MDL Number: MFCD00039288
CAS Number: 2366-52-1

Order Now

Specification Test Results

Boiling point 31.9°/745m
Density 0.7761

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR