5H-Octafluoropentanoyl chloride ≥97%

Supplier: Matrix Scientific

006500-1G
101779-136EA 63.08 USD
101779-136
5H-Octafluoropentanoyl chloride ≥97%
5H-Octafluoropentanoyl chloride

Hazard: Corrosive/Stench

Formula: C₅HClF₈O
Boiling Pt: 86…88 °C
Density: 1.67
MDL Number: MFCD00153221
CAS Number: 376-71-6

Order Now

Specification Test Results

Boiling point 86-88°
Density 1.67
ND20 1.324

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR