5H-Octafluoropentanoic acid ≥97%

Supplier: Matrix Scientific

006498-25G 006498-5G
101779-134EA 438.46 USD
101779-134 101779-132
5H-Octafluoropentanoic acid ≥97%
5H-Octafluoropentanoic acid

Hazard: Corrosive

Formula: C₅H₂F₈O₂
Boiling Pt: 165…166 °C
Melting Pt: 0 °C
Density: 1.725
MDL Number: MFCD00054690
CAS Number: 376-72-7

Order Now

Specification Test Results

Boiling point 165-166°
Density 1.725

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR