Bis(trifluoromethyl)disulfide ≥99%

Supplier: Matrix Scientific

006494-10G
101779-130EA 1869.23 USD
101779-130
Bis(trifluoromethyl)disulfide ≥99%
Bis(trifluoromethyl)disulfide

Hazard: Toxic by inhalation

Formula: C₂F₆S₂
MW: 202.14 g/mol
Boiling Pt: 34 °C
Density: 1.51
MDL Number: MFCD00041514
CAS Number: 372-64-5

Order Now

Specification Test Results

Boiling point 34°
Density 1.51

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR