3'-(Trifluoromethylthio)acetophenone

Supplier: Matrix Scientific

006487-1G
101779-124EA 80 USD
101779-124
3'-(Trifluoromethylthio)acetophenone
3'-(Trifluoromethylthio)acetophenone

Hazard: Toxic

Formula: C₉H₇F₃OS
MDL Number: MFCD00236331
CAS Number: 56773-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR