1-Ethyl-4-fluorobenzene ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-4-ethylbenzene
, 4-Ethylfluorobenzene
, (4-Fluorophenyl)ethane
006461-5G
101779-102EA
90.77
USD
101779-102
1-Ethyl-4-fluorobenzene ≥98%
1-Ethyl-4-fluorobenzene
Hazard: Flammable
Formula:
C₈H₉F MW: 124.16 g/mol Boiling Pt: 139 °C Density: 0.98 Flash Pt: 30 °C |
MDL Number:
MFCD00039218 CAS Number: 459-47-2 |
Specification Test Results
Boiling point | 139° |
Density | 0.98 |
ND20 | 1.471 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...