Methyl fluoroacetate ≥99%

Supplier: Matrix Scientific

006459-25G
101779-096EA 255.38 USD
101779-096
Methyl fluoroacetate ≥99%
Methyl fluoroacetate

Hazard: Toxic

Formula: C₃H₅FO₂
MW: 92.07 g/mol
Boiling Pt: 104 °C
Density: 1.17
MDL Number: MFCD00040096
CAS Number: 453-18-9

Order Now

Specification Test Results

Boiling point 104°
Density 1.17

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR