Oxalyl fluoride ≥97%

Supplier: Matrix Scientific

006450-25G
101779-086EA 1076.92 USD
101779-086
Oxalyl fluoride ≥97%
Oxalyl fluoride

TCA listed: T
Hazard: Toxic by inhalation/Corrosive/Nonflammable Gas

Formula: C₂F₂O₂
Boiling Pt: 26.6 °C
Melting Pt: –3 °C
MDL Number: MFCD00039219
CAS Number: 359-40-0

Order Now

Specification Test Results

Boiling point 26.6°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR