4-Fluorostyrene ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Fluoro-4-vinylbenzene
006416-100G
006416-25G
006416-5G
101779-068EA
266.8
USD
101779-068
101779-066
101779-064
4-Fluorostyrene ≥95%
4-Fluorostyrene
Hazard: Flammable/Refrigerate
Formula:
C₈H₇F MW: 122.14 g/mol Boiling Pt: 67 °C (50 mmHg) Density: 1.02 |
MDL Number:
MFCD00000361 CAS Number: 405-99-2 |
Specification Test Results
Boiling point | 67°/50mm |
Density | 1.02 |
ND20 | 1.515 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...