4-tert-Butylphenyl isothiocyanate ≥97%

Supplier: Matrix Scientific

Synonyms: 4-tert-Butylphenylisothiocyanate

006406-5G 006406-1G
101779-056EA 315.38 USD
101779-056 101779-054
4-tert-Butylphenyl isothiocyanate ≥97%
4-tert-Butylphenyl isothiocyanate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₁₁H₁₃NS
MW: 191.2926 g/mol
Boiling Pt: 275…277 °C
Melting Pt: 41…42 °C
MDL Number: MFCD00022059
CAS Number: 19241-24-8

Order Now

Specification Test Results

Boiling point 275-277°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR