2-Hydroxythiophenol ≥98%
Supplier: Matrix Scientific
Synonyms:
o-Hydroxythiophenol
, 2-Mercaptophenol
, 1-Hydroxy-2-mercaptobenzene
, 2-Hydroxyphenyl mercaptan
, 2-Hydroxybenzene-1-thiol
, 2-Hydroxybenzenethiol
006317-25G
006317-100G
101779-014EA
232.31
USD
101779-014
101779-012
2-Hydroxythiophenol ≥98%
2-Hydroxythiophenol
Hazard: Toxic/Corrosive/Air Sensitive/Stench
Formula:
C₆H₆OS MW: 126.18 g/mol Boiling Pt: 88…90 °C (8 mmHg) Density: 1.22 |
MDL Number:
MFCD00040447 CAS Number: 1121-24-0 |
Specification Test Results
Boiling point | 88-90°/8mm |
Density | 1.22 |
ND20 | 1.606 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...