Ethyl pentafluoropropionate ≥99%

Supplier: Matrix Scientific

006276-100G
101778-998EA 69 USD
101778-998
Ethyl pentafluoropropionate ≥99%
Ethyl pentafluoropropionate

TCA listed: T
Hazard: Flammable/Irritant

Formula: C₅H₅F₅O₂
MW: 192.09
Boiling Pt: 75…76 °C
Density: 1.299
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Boiling point 75-76°
Density 1.299
ND20 1.301

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR