1-Ethylpyrrole ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Ethylpyrrole
006165-25G
006165-5G
101778-922EA
176.92
USD
101778-922
101778-924
1-Ethylpyrrole ≥97%
1-Ethylpyrrole
Hazard: Flammable
Formula:
C₆H₉N MW: 95.14 g/mol Boiling Pt: 129…130 °C Density: 0.88 |
MDL Number:
MFCD00060633 CAS Number: 617-92-5 |
Specification Test Results
Boiling point | 129-130° |
Density | 0.88 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...