Tetrafluorosuccinic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3-Tetrafluorosuccinic acid
, Tetrafluoro-1,4-butanedioic acid
006154-25G
006154-5G
101778-914EA
64.62
USD
101778-914
101778-916
Tetrafluorosuccinic acid ≥98%
Tetrafluorosuccinic acid
TCA listed: T
Hazard: Corrosive/Irritant
Formula:
C₄H₂F₄O₄ MW: 190.05 g/mol Boiling Pt: 150 °C (15 mmHg) Melting Pt: 118…121 °C |
MDL Number:
MFCD00042430 CAS Number: 377-38-8 UN: 3261 ADR: 8,III |
Specification Test Results
Boiling point | 150°/15mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...