Pentafluoropropionic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
Perfluoropropionic acid
, Perfluoropropanoic acid
, 2,2,3,3,3-Pentafluoropropionic acid
, Pentafluoropropanoic acid
006131-100G
006131-25G
101778-886EA
26.15
USD
101778-886
101778-888
Pentafluoropropionic acid ≥98%
Pentafluoropropionic acid
TCA listed: T
Hazard: Corrosive/Stored under Argon/Hygroscopic
Formula:
C₃HF₅O₂ MW: 164.03 g/mol Boiling Pt: 96…97 °C Density: 1.561 |
MDL Number:
MFCD00004170 CAS Number: 422-64-0 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 96-97° |
Density | 1.561 |
ND20 | 1.284 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...