Pentafluoropropionic anhydride ≥98%

Supplier: Matrix Scientific

Synonyms: Pentafluoropropanoic anhydridePFPAPerfluoropropanoic anhydride

006130-25G 006130-5G
101778-882EA 53.85 USD
101778-882 101778-884
Pentafluoropropionic anhydride ≥98%
Pentafluoropropionic anhydride

TCA listed: T
Hazard: Corrosive/Moisture Sensitive/Refrigerate

Formula: C₆F₁₀O₃
MW: 310.05 g/mol
Boiling Pt: 69…70 °C (735 mmHg)
Density: 1.571
MDL Number: MFCD00000429
CAS Number: 356-42-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 69-70°/735
Density 1.571
ND20

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR