(±)-4-Fluoro-α-methylbenzylamine ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(4-Fluorophenyl)ethylamine
, DL-4-Fluoro-α-methylbenzylamine
006090-25G
006090-100G
101778-866EA
120
USD
101778-866
101778-868
(±)-4-Fluoro-α-methylbenzylamine ≥97%
(±)-4-Fluoro-α-methylbenzylamine
Hazard: Corrosive/Irritant
Formula:
C₈H₁₀FN MW: 139.17 g/mol Boiling Pt: 97 °C (35 mmHg) |
MDL Number:
MFCD00041323 CAS Number: 403-40-7 |
Specification Test Results
Boiling point | 97°/35mm |
ND20 | 1.5015 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...