4-(Difluoromethoxy)phenylacetonitrile

Supplier: Matrix Scientific

Synonyms: 2-(4-(Difluoromethoxy)phenyl)acetonitrile

006013-1G 006013-5G
101778-818EA 52.31 USD
101778-818 101778-820
4-(Difluoromethoxy)phenylacetonitrile
4-(Difluoromethoxy)phenylacetonitrile

Hazard: Toxic

Formula: C₉H₇F₂NO
MW: 183.1600 g/mol
MDL Number: MFCD00236242
CAS Number: 41429-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR