Pentafluoroethane ≥98%

005977-100G
101778-788EA 919.69 USD
101778-788
Pentafluoroethane ≥98%
Pentafluoroethane

TCA listed: T
Hazard: Nonflammable Gas

Formula: C₂HF₅
MW: 120.02 g/mol
Boiling Pt: –49 °C
Melting Pt: –103 °C
CAS Number: 354-33-6

Order Now

Specification Test Results

Boiling point -49°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR