Heptafluorobutyryl fluoride ≥97%

Supplier: Matrix Scientific

005974-25G
101778-782EA 433.85 USD
101778-782
Heptafluorobutyryl fluoride ≥97%
Heptafluorobutyryl fluoride

TCA listed: T
Hazard: Toxic by inhalation/Corrosive/Nonflammable Gas

Formula: C₄F₈O
Boiling Pt: 7…9 °C
MDL Number: MFCD01631693
CAS Number: 335-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR