Pentafluorobenzonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentafluorobenzonitrile

005947-25G 005947-100G
101778-760EA 27.69 USD
101778-760 101778-762
Pentafluorobenzonitrile ≥99%
Pentafluorobenzonitrile

TCA listed: T
Hazard: Toxic/Irritant

Formula: C₇F₅N
MW: 193.08 g/mol
Boiling Pt: 162…164 °C
Melting Pt: 2.4 °C
Density: 1.532
MDL Number: MFCD00001775
CAS Number: 773-82-0
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Boiling point 162-164°
Density 1.532
ND20 1.4425

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR