1-Iodoperfluoropropane ≥98%

Supplier: Matrix Scientific

Synonyms: Heptafluoro-1-iodopropane1,1,2,2,3,3,3-Heptafluoro-1-iodopropanePerfluoropropyl iodide

005897-25G
101778-736EA 307.69 USD
101778-736
1-Iodoperfluoropropane ≥98%
1-Iodoperfluoropropane

TCA listed: T
Hazard: Irritant/Light Sensitive/Store in Freezer

Formula: C₃F₇I
MW: 295.93 g/mol
Boiling Pt: 40…42 °C
Density: 2.05
MDL Number: MFCD00001061
CAS Number: 754-34-7

Order Now

Specification Test Results

Boiling point 40-42°
Density 2.1
ND20 1.3272

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR