α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Nitro-3,5-bis(trifluoromethyl)benzene

005880-100G
101778-728EA 98.9 USD
101778-728
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene ≥98%
α,α,α,α',α',α'-Hexafluoro-5-nitroxylene

Hazard: Irritant

Formula: C₈H₃F₆NO₂
MW: 259.11 g/mol
Boiling Pt: 72 °C (7 mmHg)
Density: 1.535
MDL Number: MFCD00000384
CAS Number: 328-75-6

Order Now

Specification Test Results

Boiling point 72°/7mm
Density 1.535
ND20 1.427

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR