3,5-Bis(trifluoromethyl)benzoyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3,5-Bis(trifluoromethyl)benzene-1-carbonyl chloride
, 3,5-bis(trifluoromethyl)benzoylchloride
005877-100G
005877-25G
101778-720EA
192.31
USD
101778-720
101778-718
3,5-Bis(trifluoromethyl)benzoyl chloride ≥97%
3,5-Bis(trifluoromethyl)benzoyl chloride
Hazard: Corrosive/Lachrymator
Formula:
C₉H₃ClF₆O MW: 276.57 g/mol Boiling Pt: 65…67 °C (12 mmHg) Density: 1.526 |
MDL Number:
MFCD00000387 CAS Number: 785-56-8 UN: 3265 ADR: 8,II |
Specification Test Results
Boiling point | 65-67°/12m |
Density | 1.526 |
ND20 | 1.4332 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...