3,5-Bis(trifluoromethyl)anisole

Supplier: Matrix Scientific

Synonyms: 1-Methoxy-3,5-bis(trifluoromethyl)benzene

005854-5G
101778-684EA 67.69 USD
101778-684
3,5-Bis(trifluoromethyl)anisole
3,5-Bis(trifluoromethyl)anisole

Hazard: Flammable

Formula: C₉H₆F₆O
MW: 244.14 g/mol
Boiling Pt: 158…160 °C
Flash Pt: 55 °C
MDL Number: MFCD00728736
CAS Number: 349-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR