2,6-Bis(trifluoromethyl)benzoyl chloride ≥98%

Supplier: Matrix Scientific

005826-5G 005826-1G
101778-660EA 403.08 USD
101778-660 101778-658
2,6-Bis(trifluoromethyl)benzoyl chloride ≥98%
2,6-Bis(trifluoromethyl)benzoyl chloride

Hazard: Corrosive/Moisture Sensitive

Formula: C₉H₃ClF₆O
MW: 276.57 g/mol
MDL Number: MFCD00061228
CAS Number: 53130-44-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR