2,4-Bis(trifluoromethyl)benzoyl chloride ≥98%
Supplier: Matrix Scientific
005825-1G
005825-5G
101778-654EA
58.46
USD
101778-654
101778-656
2,4-Bis(trifluoromethyl)benzoyl chloride ≥98%
2,4-Bis(trifluoromethyl)benzoyl chloride
Hazard: Corrosive
Formula:
C₉H₃ClF₆O MW: 276.57 g/mol Boiling Pt: 191…193 °C |
MDL Number:
MFCD00061227 CAS Number: 53130-43-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...