2,4-Bis(trifluoromethyl)benzonitrile ≥98%
Supplier: Matrix Scientific
005823-1G
005823-5G
101778-650EA
152.31
USD
101778-650
101778-652
2,4-Bis(trifluoromethyl)benzonitrile ≥98%
2,4-Bis(trifluoromethyl)benzonitrile
Hazard: Toxic
Formula:
C₉H₃F₆N MW: 239.12 g/mol |
MDL Number:
MFCD00061225 CAS Number: 177952-38-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...