n-Butyl trifluoroacetate ≥97%

Supplier: Matrix Scientific

005804-25G 005804-5G
101778-630EA 33.85 USD
101778-630 101778-632
n-Butyl trifluoroacetate ≥97%
n-Butyl trifluoroacetate

Hazard: Flammable

Formula: C₆H₉F₃O₂
MW: 170.13 g/mol
Boiling Pt: 100 °C
Density: 1.027
Flash Pt: 15 °C
MDL Number: MFCD00039234
CAS Number: 367-64-6

Order Now

Specification Test Results

Boiling point 100°
Density 1.027
ND20 1.353

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR