N,O-Bis(trifluoroacetyl)hydroxylamine ≥97%
Supplier: Matrix Scientific
005802-5G
101778-628EA
438.46
USD
101778-628
N,O-Bis(trifluoroacetyl)hydroxylamine ≥97%
N,O-Bis(trifluoroacetyl)hydroxylamine
Purity: 97%
CAS: 684-78-6
MDL Number: MFCD00013565
Molecular Formula: C4HF6NO3
Molecular Weight: 225,05
Hazard: Corrosive
Formula:
C₄HF₆NO₃ Melting Pt: 59…60 °C |
MDL Number:
MFCD00013565 CAS Number: 684-78-6 |
Specification Test Results
Melting point | 59-60° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...