Triisopropyl borate ≥99%

Synonyms: Tri-i-propylborate

005755-2.5KG
101778-616EA 201.46 USD
101778-616
Triisopropyl borate ≥99%
Triisopropyl borate

TCA listed: T
Hazard: Flammable/Irritant

Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139…141 °C
Density: 0.815
MDL Number: MFCD00008872
CAS Number: 5419-55-6
UN: 2616
ADR: 3,II

Order Now

Specification Test Results

Boiling point 139-141°
Density 0.815

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR