4-Methoxyphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

005703-5G 005703-25G
101778-594EA 326.15 USD
101778-594 101778-592
4-Methoxyphenyl isothiocyanate ≥98%
4-Methoxyphenyl isothiocyanate

Hazard: Corrosive/Moisture Sensitive/Lachrymator

Formula: C₈H₇NOS
MW: 165.22 g/mol
Boiling Pt: 280…281 °C
Melting Pt: 18 °C
Density: 1.196
MDL Number: MFCD00011676
CAS Number: 2284-20-0

Order Now

Specification Test Results

Boiling point 280-281°
Density 1.196
ND20 1.6489

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR