4-Phenoxyphenyl isothiocyanate ≥98%

Supplier: Matrix Scientific

005700-5G
101778-586EA 76.92 USD
101778-586
4-Phenoxyphenyl isothiocyanate ≥98%
4-Phenoxyphenyl isothiocyanate

Hazard: Toxic/Refrigerate/Lachrymator

Formula: C₁₃H₉NOS
Melting Pt: 42 °C
MDL Number: MFCD00060376
CAS Number: 3529-87-1

Order Now

Specification Test Results

Melting point 42°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR