Perfluorotoluene ≥98%
Supplier: Matrix Scientific
Synonyms:
Octafluorotoluene
, 1,2,3,4,5-Pentafluoro-6-(trifluoromethyl)benzene
, α,α,α,2,3,4,5,6-Octafluorotoluene
, Pentafluoro(trifluoromethyl)benzene
005644-100G
101778-578EA
460
USD
101778-578
Perfluorotoluene ≥98%
Perfluorotoluene
TCA listed: T
Hazard: Flammable
Formula:
C₇F₈ MW: 236.06 g/mol Boiling Pt: 104 °C Density: 1.666 |
MDL Number:
MFCD00000375 CAS Number: 434-64-0 UN: 1992 ADR: 3,II |
Specification Test Results
Boiling point | 104° |
Density | 1.666 |
ND20 | 1.368 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...