3,4-Difluorotoluene ≥98%

Supplier: Matrix Scientific

005628-25G 005628-5G
101778-556EA 36.92 USD
101778-556 101778-558
3,4-Difluorotoluene ≥98%
3,4-Difluorotoluene

Hazard: Flammable

Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 110…113 °C
Density: 1.12
Flash Pt: 25 °C
MDL Number: MFCD00075087
CAS Number: 2927-34-6
UN: 2388
ADR: 3,II

Order Now

Specification Test Results

Boiling point 110-113°
Density 1.12
ND20 1.4489

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR